Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106872 Supplier Invitrogen™ Supplier No. PA5106872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNE1.

Kcne1 is a member of Voltage-gated potassium (Kv) channels that represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. The diverse functions of Kv channels include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The Kcne1 gene encodes a membrane protein which has sequence similarity to the KCNE1 gene product, a member of the potassium channel, voltage-gated, isk-related subfamily. Kcne1, an intronless gene is deleted in AMME contiguous gene syndrome and may be involved in the cardiac and neurologic abnormalities found in the AMME contiguous gene syndrome. Functionally, Kcne1 is a potassium channel ancillary subunit that is essential for generation of some native K(+) currents by virtue of formation of heteromeric ion channel complex with voltage-gated potassium (Kv) channel pore-forming alpha subunits. Kcne1 also functions as an inhibitory beta-subunit of the repolarizing cardiac potassium ion channel KCNQ1. Diseases associated with KCNE1 include Jervell And Lange-Nielsen Syndrome 2 and Long Qt Syndrome 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNE1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Kcne1
Gene Accession No. P15382, P15383, P23299
Gene Alias cardiac delayed rectifier potassium channel protein; delayed rectifier potassium channel subunit IsK; IKs producing slow voltage-gated potassium channel subunit beta Mink; Isk; JLNS; JLNS 2; JLNS2; KCNE 1; Kcne1; KCNE1 (IsK); LQT 5; LQT2/5; LQT5; m; MGC33114; Minimal potassium channel; mink; mISK; nmf190; potassium (K+) channel protein, slowly activating (Isk); potassium channel, voltage gated subfamily E regulatory beta subunit 1; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 1; potassium voltage-gated channel Shaw-related subfamily member 1; potassium voltage-gated channel subfamily E member 1; potassium voltage-gated channel subfamily E member 1-like; potassium voltage-gated channel subfamily E regulatory subunit 1; potassium voltage-gated channel, Isk-related family, member 1; potassium voltage-gated channel, Isk-related subfamily, member 1; voltage gated potassiun channel accessory subunit
Gene Symbols Kcne1
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNE1(Accession P15382), corresponding to amino acid residues F54-R104.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16509, 25471, 3753
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.