Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNE1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | KCNE1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
KCNE1 Polyclonal specifically detects KCNE1 in Human, Mouse samples. It is validated for Western Blot.Specifications
KCNE1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3753 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Delayed rectifier potassium channel subunit IsK, FLJ18426, FLJ38123, IKs producing slow voltage-gated potassium channel subunit beta Mink, ISK, Isk-related subfamily, member 1, JLNS2FLJ94103, LQT2/5, MGC33114, Minimal potassium channel, MinK, potassium voltage-gated channel, Isk-related family, member 1, voltage gated potassiun channel accessory subunit | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KCNE1 (NP_000210.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title