Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118799 Supplier Invitrogen™ Supplier No. PA5118799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNE2.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, isk-related subfamily. This member is a small integral membrane subunit that assembles with the KCNH2 gene product, a pore-forming protein, to alter its function. This gene is expressed in heart and muscle and the gene mutations are associated with cardiac arrhythmia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNE2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNE2
Gene Accession No. P63161, Q9D808, Q9Y6J6
Gene Alias 2200002I16Rik; ATFB4; AW048273; cardiac voltage-gated potassium channel accessory subunit 2; CDW210B; CRF2-4; CRFB4; D21S58; D21S66; IL-10R2; Kcne2; LQT5; LQT6; minimum potassium ion channel related peptide 1; minimum potassium ion channel-related peptide 1; MinK-related peptide 1; minK-related peptide-1; MIRP1; Potassium channel subunit beta MiRP1; potassium channel subunit, MiRP1; potassium channel, voltage gated subfamily E regulatory beta subunit 2; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2; potassium voltage-gated channel subfamily E member 2; potassium voltage-gated channel subfamily E regulatory subunit 2; potassium voltage-gated channel, Isk-related family, member 2; potassium voltage-gated channel, Isk-related subfamily, gene 2; voltage-gated K+ channel subunit MIRP1
Gene Symbols KCNE2
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNE2(Accession Q9Y6J6), corresponding to amino acid residues Y52-N102.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171138, 246133, 9992
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.