Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533865
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533865 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533865 Supplier Invitrogen™ Supplier No. PA533865
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNE3 Polyclonal Antibody for IHC (P)

Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. Associated with KCNC4/Kv3.4 is proposed to form the subthreshold voltage-gated potassium channel in skeletal muscle and to establish the resting membrane potential (RMP) in muscle cells. Associated with KCNQ1/KCLQT1 may form the intestinal cAMP-stimulated potassium channel involved in chloride secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNE3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Kcne3
Gene Accession No. Q9JJV7, Q9WTW2, Q9Y6H6
Gene Alias 2210017H05Rik; cardiac voltage-gated potassium channel accessory subunit; HOKPP; HYPP; KCNE3; Minimum potassium ion channel-related peptide 2; minK-related peptide 2; MiRP2; potassium channel subunit beta MiRP2; potassium channel, voltage gated subfamily E regulatory beta subunit 3; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 3; potassium voltage-gated channel subfamily E member 3; potassium voltage-gated channel subfamily E regulatory subunit 3; potassium voltage-gated channel, Isk-related family, member 3; potassium voltage-gated channel, Isk-related subfamily, gene 3; potassium voltage-gated channel, Isk-related subfamily, member 3; voltage-gated K+ channel subunit MIRP2
Gene Symbols Kcne3
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from internal region of human KCNE3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100065199, 10008, 100357883, 100755499, 397168, 428105, 485197, 527762, 57442, 63883
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Hamster, Pig, Horse, Chicken, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.