Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNE4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588404 Supplier Invitrogen™ Supplier No. PA588404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YMKSKRREKK SSLLLLYKDE ERLWGEAMKP LPVVSGLRSV QVPLMLNMLQ ESVAPALSCT LCSMEGDSVS SESSSPDVHL TIQEEGADDE LEETSETPLN ESSEGSSENI HQNS; Positive Samples: 293T, LO2; Cellular Location: Membrane, Single-pass membrane protein.

Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. May associate with KCNQ1/KVLTQ1 and inhibit potassium current.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNE4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNE4
Gene Accession No. Q8WWG9
Gene Alias AW487937; cardiac voltage-gated potassium channel accessory subunit 4; KCNE4; Minimum potassium ion channel-related peptide 3; minK-related peptide 3; MIRP3; potassium channel subunit beta MiRP3; potassium channel, voltage gated subfamily E regulatory beta subunit 4; potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 4; potassium voltage-gated channel subfamily E member 4; potassium voltage-gated channel subfamily E regulatory subunit 4; potassium voltage-gated channel, Isk-related family, member 4; potassium voltage-gated channel, Isk-related subfamily, gene 4
Gene Symbols KCNE4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 108-221 of human KCNE4 (NP_5424023).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23704
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.