Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101764
Description
Antibody detects endogenous levels of total KCNF1.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily F. This gene is intronless and expressed in all tissues tested, including the heart, skeletal muscle, brain, kidney, and pancreas.
Specifications
KCNF1 | |
Polyclonal | |
Unconjugated | |
Kcnf1 | |
Gm1182; IK8; KCNF; Kcnf1; kH1; KV5.1; potassium channel KH1; potassium channel, voltage-gated modifier subfamily F, member 1; potassium voltage-gated channel modifier subfamily F member 1; potassium voltage-gated channel subfamily F member 1; potassium voltage-gated channel, subfamily F, member 1; voltage-gated potassium channel subunit Kv5.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3754, 382571 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7TSH7, Q9H3M0 | |
Kcnf1 | |
A synthesized peptide derived from human KCNF1(Accession Q9H3M0), corresponding to amino acid residues G153-T203. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction