Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584543 Supplier Invitrogen™ Supplier No. PA584543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PICFKNEMDF WKVDLKFLDD CCKSHLSEKR EELEEIARRV QLILDDLGVD A Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily F. This gene is intronless and expressed in all tissues tested, including the heart, skeletal muscle, brain, kidney, and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kcnf1
Gene Accession No. Q9H3M0
Gene Alias Gm1182; IK8; KCNF; Kcnf1; kH1; KV5.1; potassium channel KH1; potassium channel, voltage-gated modifier subfamily F, member 1; potassium voltage-gated channel modifier subfamily F member 1; potassium voltage-gated channel subfamily F member 1; potassium voltage-gated channel, subfamily F, member 1; voltage-gated potassium channel subunit Kv5.1
Gene Symbols Kcnf1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNF1. Recombinant protein control fragment (Product #RP-108224).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3754
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.