Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559033 Supplier Thermo Scientific Supplier No. PA559033
Add to Cart
Add to Cart

KCNG1 Polyclonal Antibody

KCNG1 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNG1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNG1
Gene Accession No. Q9UIX4
Gene Alias AW536275; K13; KCNG; KCNG1; kH2; KV6.1; orthologue of H. sapiens potassium voltage-gated channel, subfamily G, member 1 (KCNG1); OTTMUSG00000016048; potassium channel KH2; potassium channel Kv6.1; potassium channel, voltage gated modifier subfamily G, member 1; potassium channel, voltage-gated modifier subfamily G, member 1; potassium voltage-gated channel modifier subfamily G member 1; potassium voltage-gated channel subfamily G member 1; potassium voltage-gated channel, subfamily G, member 1; voltage-gated potassium channel subunit Kv6.1
Gene Symbols KCNG1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNG1. Recombinant protein control fragment (Product #RP-98323).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3755
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.