Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNG2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989401
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9401 Supplier Bioss Supplier No. BS9857R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit of the voltage-gated potassium channel. The delayed-rectifier type channels containing this subunit may contribute to cardiac action potential repolarization. [provided by RefSeq, Jul 2008].

Specifications

Antigen KCNG2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KCNG2
Gene Accession No. Q9UJ96
Gene Alias AA591524; Cardiac potassium channel subunit; cardiac potassium channel subunit (Kv6.2); KCNF2; KCNG2; Kv6.2; potassium channel, voltage gated modifier subfamily G, member 2; potassium voltage-gated channel modifier subfamily G member 2; Potassium voltage-gated channel subfamily G member 2; potassium voltage-gated channel, subfamily G, member 2; Voltage-gated potassium channel subunit Kv6.2
Gene Symbols KCNG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KCNG2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26251
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.