Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106704 Supplier Invitrogen™ Supplier No. PA5106704
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNG3.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit functioning as a modulatory molecule.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNG3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNG3
Gene Accession No. P59053, Q8R523, Q8TAE7
Gene Alias KCNG3; Kv10.1; Kv10.1a; Kv10.1b; Kv6.3; potassium channel, voltage gated modifier subfamily G, member 3; potassium voltage-gated channel modifier subfamily G member 3; potassium voltage-gated channel subfamily G member 3; potassium voltage-gated channel subfamily G memeber 3; potassium voltage-gated channel, subfamily G, member 3; potassium voltage-gated channel, subfamily G, memeber 3; voltage-gated potassium channel 6.3; voltage-gated potassium channel Kv10.1; voltage-gated potassium channel subunit Kv10.1; Voltage-gated potassium channel subunit Kv6.3; voltage-gated potassium channel subunit Kv6.4
Gene Symbols KCNG3
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNG3(Accession Q8TAE7), corresponding to amino acid residues V181-W231.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170850, 171011, 225030
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.