Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528353
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528353 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528353 Supplier Invitrogen™ Supplier No. PA528353
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3. Predicted reactivity: Mouse (84%), Rat (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member functions as a modulatory subunit. The gene has strong expression in brain. Multiple alternatively spliced variants have been found in normal and cancerous tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene KCNG4
Gene Accession No. Q80XM3, Q8TDN1
Gene Alias 4921535I01Rik; AW049024; KCNG3; Kcng4; KV6.3; KV6.4; potassium channel, voltage gated modifier subfamily G, member 4; potassium voltage-gated channel modifier subfamily G member 4; potassium voltage-gated channel subfamily G member 4; potassium voltage-gated channel, subfamily G, member 4; voltage-gated potassium channel Kv6.3; voltage-gated potassium channel subunit Kv6.3; voltage-gated potassium channel subunit Kv6.4
Gene Symbols KCNG4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KCNG4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66733, 93107
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.