Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113603 Supplier Invitrogen™ Supplier No. PA5113603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELILYRKSG LPFWCLLDVI PIKNEKGEVA LFLVSHKDIS ETKNRGGPDR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.

The protein encoded by this gene is a voltage-gated potassium channel alpha subunit predominantly expressed in the forebrain. Studies in mice have found that cognitive function increases when this gene is knocked out. In humans, the encoded protein has been shown to be capable of binding glycoprotein 120 of the human immunodeficiency virus type 1 (HIV-1) envelope. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNH3
Gene Accession No. Q9ULD8
Gene Alias AU019351; Bec1; Brain-specific eag-like channel 1; C030044P22Rik; ELK channel 2; Elk2; ether a go-go like; ether-a-go-go K(+) channel family member; ether-a-go-go-like potassium channel 2; KCNH3; KIAA1282; Kv12.2; Melk2; potassium channel, voltage gated eag related subfamily H, member 3; potassium voltage-gated channel subfamily H member 3; potassium voltage-gated channel, subfamily H (eag-related), member 3; rElk2; voltage-gated potassium channel subunit Kv12.2
Gene Symbols KCNH3
Host Species Rabbit
Immunogen Synthetic peptide located within the following region: AELILYRKS GLPFWCLLD VIPIKNEKGE VALFLVSHKD ISETKNRGGP DR (aa 95-144).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23416
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.