Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNH3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539353
Description
Specifications
| KCNH3 | |
| Polyclonal | |
| Unconjugated | |
| KCNH3 | |
| Elk2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16512, 23416, 27150 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ULD8, Q9WVJ0, O89047 | |
| KCNH3 | |
| A synthetic peptide derived from the internal region of human KCNH3 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction