Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556485
Description
Immunogen sequence: GDYEVIDEVT NTIQIDSWLY QLALSIGTPY RYNTSAGIWE GGPSKDS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNH5 gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH5 is not expressed in differentiating myoblasts.
Specifications
KCNH5 | |
Polyclonal | |
Unconjugated | |
KCNH5 | |
EAG2; Ether-a-go-go potassium channel 2; ether-a-go-go-related potassium channel 2; hEAG2; H-EAG2; KCNH5; Kv10.2; potassium channel, voltage gated eag related subfamily H, member 5; potassium voltage-gated channel subfamily H member 5; potassium voltage-gated channel, subfamily H (eag-related), member 5; rEAG2; Voltage-gated potassium channel subunit Kv10.2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27133 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCM2 | |
KCNH5 | |
Recombinant protein corresponding to Human KCNH5. Recombinant protein control fragment (Product #RP-89132). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction