Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139910 Supplier Invitrogen™ Supplier No. PA5139910
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SLKQNNRDAM ELKPNGGADQ KCLKVNSPIR MKNGNGKGWL RLKNNMGAHE EKKEDWNNVT KAESMGLLSE DPKSSDSENS VTK Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 95%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNH5 gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH5 is not expressed in differentiating myoblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNH5
Gene Accession No. Q8NCM2
Gene Alias EAG2; Ether-a-go-go potassium channel 2; ether-a-go-go-related potassium channel 2; hEAG2; H-EAG2; KCNH5; Kv10.2; potassium channel, voltage gated eag related subfamily H, member 5; potassium voltage-gated channel subfamily H member 5; potassium voltage-gated channel, subfamily H (eag-related), member 5; rEAG2; Voltage-gated potassium channel subunit Kv10.2
Gene Symbols KCNH5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNH5. Recombinant protein control fragment (Product #RP-109286).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27133
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.