Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541073 Supplier Invitrogen™ Supplier No. PA541073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LTNSRSVLQQ LTPMNKTEVV HKHSRLAEVL QLGSDILPQY KQEAPKTPPH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNH5 gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH5 is not expressed in differentiating myoblasts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNH5
Gene Accession No. Q8NCM2
Gene Alias EAG2; Ether-a-go-go potassium channel 2; ether-a-go-go-related potassium channel 2; hEAG2; H-EAG2; KCNH5; Kv10.2; potassium channel, voltage gated eag related subfamily H, member 5; potassium voltage-gated channel subfamily H member 5; potassium voltage-gated channel, subfamily H (eag-related), member 5; rEAG2; Voltage-gated potassium channel subunit Kv10.2
Gene Symbols KCNH5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCNH5 (aa 156-205).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27133
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.