Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550775 Supplier Invitrogen™ Supplier No. PA550775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total KCNH6 protein.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. KCNH6 encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNH6
Gene Accession No. Q9H252
Gene Alias Eag-related gene member 2; eag-related protein 2; Erg2; ERG-2; Ether-a-go-go-related gene potassium channel 2; ether-a-go-go-related protein 2; HERG2; hERG-2; KCNH6; Kv11.2; m-erg2; potassium channel, voltage gated eag related subfamily H, member 6; potassium voltage-gated channel subfamily H member 6; potassium voltage-gated channel, subfamily H (eag-related), member 6; voltage-gated potassium channel subunit Kv11.2
Gene Symbols KCNH6
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the N terminal of human potassium voltage-gated channel, subfamily H (eag-related), member 6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81033
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.