Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNH8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568690 Supplier Invitrogen™ Supplier No. PA568690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 92%; Rat: 85%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kcnh8
Gene Accession No. P59111
Gene Alias C130090D05Rik; ELK; ELK channel 3; ELK1; Elk3; ether-a-go-go-like potassium channel 1; ether-a-go-go-like potassium channel 3; Ets-1; hElk1; Kcnh8; Kv12.1; potassium channel, voltage gated eag related subfamily H, member 8; potassium voltage-gated channel subfamily H member 8; potassium voltage-gated channel, subfamily H (eag-related), member 8; potassium voltage-gated channel, subfamily H, member 8; voltage-gated potassium channel subunit Kv12.1
Gene Symbols Kcnh8
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse KCNH8.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211468
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.