Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-110 Supplier Invitrogen™ Supplier No. PA141705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human heart and brain protein or tissue. Immunogen sequence: LQTKQRRPSK DKIEDELE.

This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% gelatin and 0.05% sodium azide
Gene Kcnip1
Gene Accession No. Q8R426, Q9JJ57, Q9NZI2
Gene Alias ABP; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; Kchip1.2; KCHIP2; Kcnip1; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; potassium channel-interacting protein 1; potassium voltage-gated channel interacting protein 1; VABP; Vesicle APC-binding protein
Gene Symbols Kcnip1
Host Species Rabbit
Immunogen Within the range of amino acids 1-35 of human ABP protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30820, 541458, 595102, 65023, 70357
Target Species Human, Mouse, Rat, Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.