Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCNIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116460
Description
Positive controls: Human skin tissue lysates, rat brain tissue, SH-SY5Y.
This gene encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. Alternative splicing results in multiple transcript variant encoding different isoforms.
Specifications
| KCNIP1 | |
| Polyclonal | |
| Unconjugated | |
| Kcnip1 | |
| ABP; A-type potassium channel modulatory protein 1; A-type potassium channel modulatory protein 1.2; KChIP1; Kchip1.2; KCHIP2; Kcnip1; Kv channel interacting protein 1; Kv channel-interacting protein 1; potassium channel interacting protein 1; potassium channel-interacting protein 1; potassium voltage-gated channel interacting protein 1; VABP; Vesicle APC-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30820, 65023 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8R426, Q9NZI2 | |
| Kcnip1 | |
| Synthetic peptide within Human KChIP1 aa 1-50. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction