Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnip2 Mouse anti-Mouse, Rat, ATTO 488, Clone: FH9, Abnova™

Catalog No. 89338921 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-921 Supplier Abnova Corporation Supplier No. MAB19417

Mouse Monoclonal Antibody.

Kcnip2 Monoclonal antibody specifically detects Kcnip2 in Mouse, Rat samples. It is validated for WB-Ti, IHC-P.

Specifications

Antigen Kcnip2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FH9
Conjugate Atto 488
Description Kv channel-interacting protein 2
Gene Kcnip2
Gene Alias KCHIP2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Kcnip2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 56817
Target Species Mouse, Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.