Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnip2, Mouse anti-Rat, Biotin, Clone: FH9, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16521
Specifications
Kcnip2 | |
Monoclonal | |
Biotin | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
KCHIP2 | |
Mouse | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
FH9 | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Kv channel-interacting protein 2 | |
Kcnip2 | |
Recombinant protein corresponding to full length rat Kcnip2. | |
RUO | |
56817 | |
Store at 4°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction