Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnip3 Mouse anti-Rat, Alkaline Phosphatase, Clone: X12, Abnova™

Catalog No. 89339002 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-002 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-002 Supplier Abnova Corporation Supplier No. MAB19717

Mouse Monoclonal Antibody.

Kcnip3 Monoclonal antibody specifically detects Kcnip3 in Rat samples. It is validated for WB, IHC-P.

Specifications

Antigen Kcnip3
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone X12
Conjugate Alkaline Phosphatase (AP)
Description Kv channel interacting protein 3, calsenilin
Gene Kcnip3
Gene Alias Csen, Dream, rKChIP3
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of rat Kcnip3
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 65199
Target Species Rat
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.