Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kcnip3 Mouse anti-Rat, ATTO 390, Clone: X12, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB19709
Description
Kcnip3 Monoclonal antibody specifically detects Kcnip3 in Rat samples. It is validated for WB, IHC-P.Specifications
Kcnip3 | |
Monoclonal | |
Atto 390 | |
Kcnip3 | |
Mouse | |
100 μg | |
65199 | |
Store at 4°C | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
X12 | |
Kv channel interacting protein 3, calsenilin | |
Csen, Dream, rKChIP3 | |
Recombinant protein corresponding to full length of rat Kcnip3 | |
Primary | |
Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction