Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNIP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540940 Supplier Invitrogen™ Supplier No. PA540940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NSTKRSIKER LMKLLPCSAA KTSSPAIQNS VEDELEMATV RHRPEALELL Sequence homology: Cow: 93%; Dog: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNIP4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNIP4
Gene Accession No. Q6PIL6
Gene Alias A-type potassium channel modulatory protein 4; AV032399; Calp; Calp250; calsenilin-like protein; KChIP4; KChIP4a; KCNIP4; Kv channel interacting protein 4; Kv channel-interacting protein 4; potassium channel interacting protein 4; potassium channel-interacting protein 4; potassium voltage-gated channel interacting protein 4; Unknown (protein for MGC:137737)
Gene Symbols KCNIP4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCNIP4 (aa 27-76).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80333
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.