Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kir7.1 (KCNJ13) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548366
Description
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ13 has a very low single channel conductance, low sensitivity to block by external barium and cesium, and no dependence of its inward rectification properties on the internal blocking particle magnesium.
Specifications
Kir7.1 (KCNJ13) | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O60928 | |
KCNJ13 | |
KLH conjugated synthetic peptide between 67-95 amino acids from the N-terminal region of human KCNJ13. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
KCNJ13 | |
Inward rectifier potassium channel 13, Inward rectifier K(+) channel Kir71, Potassium channel, inwardly rectifying subfamily J member 13, KCNJ13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3769 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction