Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNJ14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567198 Supplier Invitrogen™ Supplier No. PA567198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETEEGNGVETE DGAASPRVLT PTLALTLPP Highest antigen sequence identity to the following orthologs - mouse 67%, rat 70%.

Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ14 gives rise to low-conductance channels with a low affinity to the channel blockers Barium and Cesium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNJ14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNJ14
Gene Accession No. Q9UNX9
Gene Alias A930026G01Rik; ATP-sensitive inward rectifier potassium channel 14; Inward rectifier K(+) channel Kir2.4; inwardly rectifying potassium channel KIR2.4; IRK4; IRK-4; KCNJ14; KIR2.4; potassium channel, inwardly rectifying subfamily J member 14; potassium channel, inwardly rectifying subfamily J, member 14; potassium inwardly-rectifying channel J14; potassium inwardly-rectifying channel, subfamily J, member 14; potassium voltage-gated channel subfamily J member 14
Gene Symbols KCNJ14
Host Species Rabbit
Immunogen Recombinant Human KCNJ14. Recombinant protein control fragment (Product #RP-105545).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3770
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.