Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kir4.2 (KCNJ15) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548818
Description
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
Specifications
Kir4.2 (KCNJ15) | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q99712 | |
KCNJ15 | |
KLH conjugated synthetic peptide between 339-367 amino acids from the C-terminal region of human KCNJ15. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KCNJ15 | |
ATP-sensitive inward rectifier potassium channel 15, Inward rectifier K(+) channel Kir13, Inward rectifier K(+) channel Kir42, Potassium channel, inwardly rectifying subfamily J member 15, KCNJ15, KCNJ14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3772 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction