Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK 9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986918
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6918 Supplier Bioss Supplier No. BS5933R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].

Specifications

Antigen KCNK 9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Kcnk1
Gene Accession No. O00180
Gene Alias AI788889; BOS_24011; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; KCNK1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; Potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1
Gene Symbols Kcnk1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KCNK9.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3775
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.