Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541010 Supplier Invitrogen™ Supplier No. PA541010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EDQVHIIEHD QLSFSSITDQ AAGMKEDQKQ NEPFVATQSS ACVDGPANH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 93%.

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kcnk1
Gene Accession No. O00180
Gene Alias AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788)
Gene Symbols Kcnk1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KCNK1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3775
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.