Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572833
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572833 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572833 Supplier Invitrogen™ Supplier No. PA572833
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNK1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Kcnk1
Gene Accession No. O00180, O08581, Q9Z2T2
Gene Alias AI788889; double-pore potassium channel; DPK; HOHO; HOHO1; Inward rectifying potassium channel protein TWIK-1; K2P1; K2p1.1; Kcnk1; kcnk1 {ECO:0000312; KCNO1; Potassium channel K2P1; potassium channel KCNO1; Potassium channel subfamily K member 1; potassium channel TWIK-1; potassium channel, subfamily K, member 1; potassium channel, two pore domain subfamily K, member 1; potassium inwardly-rectifying channel, subfamily K, member 1; potassium two pore domain channel subfamily K member 1; putative potassium channel TWIK; rabKCNK1; RGD:621447}; rTWIK; tandem of P domains in a weak inward rectifying K+ channel 1; Twik; TWIK1; TWIK-1; two-pore potassium channel; Unknown (protein for MGC:142788)
Gene Symbols Kcnk1
Host Species Rabbit
Immunogen KCNK1 antibody for a 17 amino acid peptide near the carboxy terminus of human KCNK1. The immunogen is within amino acids 220 - 270 of KCNK1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16525, 3775, 59324
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.