Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588209 Supplier Invitrogen™ Supplier No. PA588209
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLGLDQRAHS LDMLSPEKRS VFAALDTGRF KASSQESINN RPNNLRLKGP EQLNKHGQGA SEDNIINKFG STSRLTKRKN KDLKKTLPED VQKIYKTFRN YSLDEEKKEE ETEKMCNSDN SSTAMLTDCI QQHAELENGM IPTDTKDREP ENNSLLEDRN; Positive Samples: Mouse spleen, Rat testis; Cellular Location: Membrane, Multi-pass membrane protein.

KCNK10 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The message for this gene is highly expressed in the kidney and pancreas. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. The protein is stimulated strongly by arachidonic acid and to a lesser degree by membrane stretching, intracellular acidification, and general anaesthetics. Three transcript variants have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNK10
Gene Accession No. P57789, Q9JIS4
Gene Alias 1700024D23Rik; 2P domain potassium channel TREK2; 3010005K24Rik; K2p10.1; Kcnk10; Outward rectifying potassium channel protein TREK-2; outward rectifying potassium channel TREK2; potassium channel subfamily K member 10; potassium channel TREK-2; potassium channel, subfamily K, member 10; potassium channel, two pore domain subfamily K, member 10; potassium two pore domain channel subfamily K member 10; PPP1R97; protein phosphatase 1, regulatory subunit 97; TREK2; TREK-2; TREK-2 K(+) channel subunit; TREK-2 two-pore-domain K+ channel; TWIK-related K+ channel 2
Gene Symbols KCNK10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 384-543 of human KCNK10 (NP_6121911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54207, 65272, 72258
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.