Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572687
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572687 Supplier Invitrogen™ Supplier No. PA572687
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KCNK12 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK12
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KCNK12
Gene Accession No. Q9ERS1, Q9HB15
Gene Alias K2p12.1; KCNK12; mntk1; Potassium channel subfamily K member 12; potassium channel, subfamily K, member 12; potassium channel, two pore domain subfamily K, member 12; potassium two pore domain channel subfamily K member 12; tandem pore domain halothane-inhibited potassium channel 2; tandem pore domain potassium channel THIK-2; THIK2; THIK-2
Gene Symbols KCNK12
Host Species Rabbit
Immunogen KCNK12 antibody for a 14 amino acid synthetic peptide near the carboxy terminus of human KCNK12. The immunogen is within amino acids 320 - 370 of KCNK12.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210741, 56660, 64119
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.