Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101758
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101758 Supplier Invitrogen™ Supplier No. PA5101758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNK12.

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK12
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNK12
Gene Accession No. Q9ERS1, Q9HB15
Gene Alias K2p12.1; KCNK12; mntk1; Potassium channel subfamily K member 12; potassium channel, subfamily K, member 12; potassium channel, two pore domain subfamily K, member 12; potassium two pore domain channel subfamily K member 12; tandem pore domain halothane-inhibited potassium channel 2; tandem pore domain potassium channel THIK-2; THIK2; THIK-2
Gene Symbols KCNK12
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNK12(Accession Q9HB15), corresponding to amino acid residues L41-R91.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210741, 56660, 64119
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.