Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK13 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen KCNK13
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP180083
SDP
View Documents
Novus Biologicals
NBP180083
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

KCNK13 Polyclonal specifically detects KCNK13 in Human samples. It is validated for Western Blot.
Specifications

Specifications

KCNK13
Polyclonal
Purified
RUO
K2p13.1, potassium channel subfamily K member 13, potassium channel, subfamily K, member 13, tandem pore domain potassium channel THIK-1, THIK-1
KCNK13
IgG
Protein A purified
Western Blot
Unconjugated
Rabbit
NP_071337
56659
Synthetic peptide directed towards the C terminal of human KCNK13. Peptide sequence SMKDLLAANKASLAILQKQLSEMANGCPHQTSTLARDNEFSGGVGAFAIM.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.