Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109876 Supplier Invitrogen™ Supplier No. PA5109876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IGFGDFVALQ SGEALQRKLP YVAFSFLYIL LGLTVIGAFL NLVVLRFLVA SADWPERAAR TPSPRPPGAP ESRGLWLPRR PARSVGSASV FCHVHKLERC A.

This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The product of this gene has not been shown to be a functional channel, however, it may require other non-pore-forming proteins for activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KCNK15
Gene Accession No. Q9H427
Gene Alias Acid-sensitive potassium channel protein TASK-5; dJ781B1.1; K2p15.1; KCNK11; KCNK14; Kcnk15; KT3.3; Potassium channel subfamily K member 15; potassium channel subfamily K member 15 (TASK-5); potassium channel, subfamily K, member 14; potassium channel, subfamily K, member 15; potassium channel, subfamily K, member 15 (TASK-5); potassium channel, two pore domain subfamily K, member 15; potassium two pore domain channel subfamily K member 15; rTASK-5; TASK5; TASK-5; TWIK-related acid-sensitive K(+) channel 5; two pore K(+) channel KT3.3; Two pore potassium channel KT3.3
Gene Symbols KCNK15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human KCNK15 (NP_071753.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60598
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.