Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560543 Supplier Invitrogen™ Supplier No. PA560543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSCCHHSSKE DFKSQSWRQG PDREPESHSP QQGCYPEGPM GIIQHLEPSA HAAGCGKDS Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 31%.

The protein encoded by this gene belongs to the family of potassium channel proteins containing two pore-forming P domains. This channel is an open rectifier which primarily passes outward current under physiological K+ concentrations. This gene is activated at alkaline pH. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNK17
Gene Accession No. Q96T54
Gene Alias 2P domain potassium channel Talk-2; Acid-sensitive potassium channel protein TASK-4; K2p17.1; KCNK17; Potassium channel subfamily K member 17; potassium channel, two pore domain subfamily K, member 17; potassium two pore domain channel subfamily K member 17; TALK2; TALK-2; TASK4; TASK-4; TWIK-related acid-sensitive K(+) channel 4; TWIK-related alkaline pH-activated K(+) channel 2; UNQ5816/PRO19634
Gene Symbols KCNK17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNK17. Recombinant protein control fragment (Product #RP-94808).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.