Learn More
Invitrogen™ KCNK18 Polyclonal Antibody

Description
A suggested positive control is rat brain tissue lysate. PA5-20873 can be used with blocking peptide PEP-0987.
Specifications
Specifications
| Antigen | KCNK18 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | KCNK18 |
| Gene Accession No. | Q6Q1P3, Q6VV64, Q7Z418 |
| Gene Alias | 2P K ion channel TRESK; Gm781; K2p18.1; Kcnk18; MGR13; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; potassium two pore domain channel subfamily K member 18; tandem-pore K+ (K(2P)) channel; Tresk; TRESK2; Tresk-2; TRIK; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel; Two-pore-domain potassium channel TRESK |
| Gene Symbols | KCNK18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.