Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNK18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA560810
Description
Immunogen sequence: ILSTSYNRFR KFPFFTRPLL SKWCPKSLFK KKPDPKPADE AVPQIIISAE ELPGPKLGTC PSR Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 48%.
KCNK18 Polyclonal antibody specifically detects KCNK18 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications
KCNK18 | |
Polyclonal | |
Unconjugated | |
KCNK18 | |
2P K ion channel TRESK; Gm781; K2p18.1; Kcnk18; MGR13; Potassium channel subfamily K member 18; potassium channel, subfamily K, member 18; potassium channel, two pore domain subfamily K, member 18; potassium two pore domain channel subfamily K member 18; tandem-pore K+ (K(2P)) channel; Tresk; TRESK2; Tresk-2; TRIK; TWIK-related individual K+ channel; TWIK-related individual potassium channel; TWIK-related spinal cord K+ channel; TWIK-related spinal cord potassium channel; Two-pore-domain potassium channel TRESK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
338567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z418 | |
KCNK18 | |
Recombinant protein corresponding to Human KCNK18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction