Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK18, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89998484 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-484 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-484 Supplier Abnova Corporation Supplier No. PAB31501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence: ILSTSYNRFRKFPFFTRPLLSKWCPKSLFKKKPDPKPADEAVPQIIISAEELPGPKLGTCPSR

Specifications

Antigen KCNK18
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Gene potassium channel, subfamily K, member 18
Gene Alias K2p18.1/TRESK/TRESK-2/TRESK2/TRIK
Gene Symbols KCNK18
Host Species Rabbit
Immunogen Recombinant protein corresponding to human KCNK18.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338567
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.