Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK3 Antibody, Novus Biologicals™
SDP

Catalog No. NB176808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176808 Supplier Novus Biologicals Supplier No. NBP313121
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNK3 Polyclonal antibody specifically detects KCNK3 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen KCNK3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias Acid-sensitive potassium channel protein TASK-1, cardiac potassium channel, K2p3.1, OAT1, potassium channel subfamily K member 3, potassium channel, subfamily K, member 3, potassium inwardly-rectifying channel, subfamily K, member 3, TASK1, TASK-1, TASKacid-sensitive potassium channel protein TASK, TBAK1, TWIK-related acid-sensitive K(+) channel 1, TWIK-related acid-sensitive K+ channel, two P domain potassium channel, Two pore K(+) channel KT3.1, Two pore potassium channel KT3.1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human KCNK3. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3777
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.