Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103173 Supplier Invitrogen™ Supplier No. PA5103173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TASK-1.

KCNK3 encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The gene product is an outwardly rectifying channel that is sensitive to changes in extracellular pH and is inhibited by extracellular acidification. Also referred to as an acid-sensitive potassium channel, it is activated by the anesthetics halothane and isoflurane. Although three transcripts are detected in northern blots, there is currently no sequence available to confirm transcript variants for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KCNK3
Gene Accession No. O14649, O35111, O54912
Gene Alias acid-sensitive potassium channel protein TASK; acid-sensitive potassium channel protein TASK-1; cardiac potassium channel; cardiac two pore background K(+) channel; Ctbak; cTBAK 1; cTBAK-1; K2p3.1; KCNK3; KCNK9; LOW QUALITY PROTEIN: potassium channel subfamily K member 3; OAT1; Potassium channel subfamily K member 3; potassium channel subfamily K member 3; LOW QUALITY PROTEIN: potassium channel subfamily K member 3; potassium channel, subfamily K, member 3; potassium channel, two pore domain subfamily K, member 3; potassium inwardly-rectifying channel, subfamily K, member 3; potassium two pore domain channel subfamily K member 3; PPH4; rTASK; TASK; TASK 1; TASK1; TASK-1; TASK-1 ion channel receptor; TASK-related potassium channel; TBAK1; TWIK-related acid-sensitive K(+) channel 1; TWIK-related acid-sensitive K+ channel; two P domain potassium channel; two pore K(+) channel KT3.1; Two pore potassium channel KT3.1
Gene Symbols KCNK3
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNK3(Accession O14649), corresponding to amino acid residues L232-T282.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16527, 29553, 3777
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.