Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNK4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169621
Description
KCNK4 Polyclonal specifically detects KCNK4 in Human samples. It is validated for Western Blot.Specifications
KCNK4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
K2p4.1, K2P4.1 potassium channel, potassium channel subfamily K member 4, potassium channel, subfamily K, member 4, TRAAKTRAAK1, TWIK-related arachidonic acid-stimulated potassium channel protein, Two pore K(+) channel KT4.1, two pore K+ channel KT4.1, Two pore potassium channel KT4.1 | |
Rabbit | |
43 kDa | |
100 μL | |
Primary | |
Canine: 86%; Equine: 86%; Bovine: 86%. | |
Human, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9NYG8 | |
KCNK4 | |
Synthetic peptides corresponding to KCNK4(potassium channel, subfamily K, member 4) The peptide sequence was selected from the N terminal of KCNK4. Peptide sequence ELGEVREKFLRAHPCVSDQELGLLIKEVADALGGGADPETNSTSNSSHSA. | |
Affinity purified | |
RUO | |
50801 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction