Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109267 Supplier Invitrogen™ Supplier No. PA5109267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Mouse colon tissue, rat kidney tissue, rat large intestine tissue, HepG2.

Potassium channels play a role in many cellular processes including maintenance of the action potential, muscle contraction, hormone secretion, osmotic regulation, and ion flow. This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. The encoded protein homodimerizes and functions as an outwardly rectifying channel. It is expressed primarily in neural tissues and is stimulated by membrane stretch and polyunsaturated fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNK4
Gene Accession No. Q9NYG8
Gene Alias K2p4.1; K2P4.1 potassium channel; Kcnk4; KT4.1; KT4.1b; LOW QUALITY PROTEIN: potassium channel subfamily K member 4; MLZ-622; Potassium channel subfamily K member 4; potassium channel, subfamily K, member 4; potassium channel, two pore domain subfamily K, member 4; potassium inwardly-rectifying channel subfamily K member 4; potassium inwardly-rectifying channel, subfamily K, member 4; potassium two pore domain channel subfamily K member 4; protein expressed in male leptotene and zygotene spermatocytes 622; Tex40; TRAAK; TRAAK1; TRAAKt; TWIK-related arachidonic acid-stimulated potassium channel protein; Two pore K(+) channel KT4.1; two pore K+ channel KT4.1; two pore potassium channel KT4.1
Gene Symbols KCNK4
Host Species Rabbit
Immunogen Synthetic peptide within Human K2P41 aa 326-375.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50801
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.