Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115453
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115453 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115453 Supplier Invitrogen™ Supplier No. PA5115453
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNK5.

KCNK5 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. KCNK5 is mainly expressed in the cortical distal tubules and collecting ducts of the kidney. The protein is highly sensitive to external pH and this, in combination with its expression pattern, suggests it may play an important role in renal potassium transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene KCNK5
Gene Accession No. O95279
Gene Alias Acid-sensitive potassium channel protein TASK-2; K2p5.1; K2P5.1 potassium channel; Kcnk5; KCNK5b; Potassium channel subfamily K member 5; potassium channel, subfamily K, member 1 (TASK-2); potassium channel, subfamily K, member 5; potassium channel, subfamily, member 5 (KCNK5); potassium channel, two pore domain subfamily K, member 5; potassium inwardly-rectifying channel, subfamily K, member 5; potassium two pore domain channel subfamily K member 5; TASK2; TASK-2; TWIK-related acid-sensitive K(+) channel 2; TWIK-related acid-sensitive K+ channel 2; two-pore potassium channel
Gene Symbols KCNK5
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNK5(Accession O95279), corresponding to amino acid residues E463-T499.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16529, 364241, 8645
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.