Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541330 Supplier Invitrogen™ Supplier No. PA541330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RLGRVVLANA SGSANASDPA WDFASALFFA STLITTVGYG YTTPLTDAGK Sequence homology: Cow: 100%; Human: 100%; Pig: 100%.

KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KCNK6
Gene Accession No. Q9Y257
Gene Alias D7Ertd764e; FLJ12282; inward rectifying potassium channel protein TWIK-2; K2p6.1; K2P6.1 potassium channel; Kcnk6; KCNK8; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; potassium two pore domain channel subfamily K member 6; TOSS; TWIK2; TWIK-2; TWIK-2 two-pore-domain K+ channel; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence
Gene Symbols KCNK6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KCNK6 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9424
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.