Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119866
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119866 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119866 Supplier Invitrogen™ Supplier No. PA5119866
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Jurkat cell lysate, rat ovary tissue lysate, human kidney tissue, human placenta tissue, mouse small intestine tissue, rat brain tissue. Subcellular Location: Membrane.

KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNK6
Gene Accession No. Q3TBV4, Q9Y257
Gene Alias D7Ertd764e; FLJ12282; inward rectifying potassium channel protein TWIK-2; K2p6.1; K2P6.1 potassium channel; Kcnk6; KCNK8; Potassium channel subfamily K member 6; potassium channel, subfamily K, member 6; potassium channel, two pore domain subfamily K, member 6; potassium inwardly-rectifying channel, subfamily K, member 6; potassium two pore domain channel subfamily K member 6; TOSS; TWIK2; TWIK-2; TWIK-2 two-pore-domain K+ channel; TWIK-originated similarity sequence; TWIK-originated sodium similarity sequence
Gene Symbols KCNK6
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal human KCNK6 from aa 291-aa313.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116491, 52150, 9424
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.