Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567461 Supplier Thermo Scientific Supplier No. PA567461
Add to Cart
Add to Cart

KCNK7 Polyclonal Antibody

KCNK7 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNK7
Gene Accession No. Q9Y2U2
Gene Alias K2p7.1; KCNK7; Potassium channel subfamily K member 7; potassium channel, subfamily K, member 7; potassium channel, two pore domain subfamily K, member 7; potassium two pore domain channel subfamily K member 7; RGD1565025; TWIK3; two pore domain K+ channel
Gene Symbols KCNK7
Host Species Rabbit
Immunogen Recombinant Human KCNK7. Recombinant protein control fragment (Product #RP-108118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.