Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK9 Mouse anti-Human, Clone: Y4, MilliporeSigma™

Catalog No. MABC1743100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1743100 100 μL
MABC174325 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1743100 Supplier MilliporeSigma Supplier No. MABC1743100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KCNK9 Monoclonal antibody specifically detects KCNK9 in Human samples. It is validated for Flow Cytometry, Function Assay, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK9
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot
Classification Monoclonal
Clone Y4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide
Host Species Mouse
Immunogen Recombinant fragment corresponding to the 59 amino acids from the first extracellular domain (M1P1 loop) of human Potassium channel subfamily K member 9 (KCNK9).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.