Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KCNK9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115454 Supplier Invitrogen™ Supplier No. PA5115454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KCNK9.

KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Kcnk9
Gene Accession No. Q3LS21, Q9ES08, Q9NPC2
Gene Alias Acid-sensitive potassium channel protein TASK-3; K2p9.1; Kcnk9; KT3.2; MGC138268; MGC138270; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel TASK3; potassium channel, subfamily K, member 9; potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; potassium two pore domain channel subfamily K member 9; TASK3; TASK-3; TWIK-related acid-sensitive K(+) channel 3; two pore K(+) channel KT3.2; Two pore potassium channel KT3.2
Gene Symbols Kcnk9
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNK9(Accession Q9NPC2), corresponding to amino acid residues I48-H98.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223604, 51305, 84429
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.